Anti-PRL3 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Lieferant: United States Biological
USBIP9109-48C3-PEEA 852 EUR
USBIP9109-48C3-PE
Anti-PRL3 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-1:500
Do not freeze!
Type: Primary
Antigen: PRL3
Clonality: polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse