Anti-Prothrombin Rat monoclonal antibody [clone: 9L609]
USBIP9115-08BEA 392 EUR
USBIP9115-08B
Anti-Prothrombin Rat monoclonal antibody [clone: 9L609]
Type: Primary
Antigen: Prothrombin
Clonality: monoclonal
Clone: 9L609
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse