Anti-Prothrombin Rat Monoclonal Antibody [clone: 11C175]
Lieferant: United States Biological
USBIP9115-17EA 405 EUR
USBIP9115-17
Anti-Prothrombin Rat Monoclonal Antibody [clone: 11C175]
Antikörper
Type: Primary
Antigen: Prothrombin
Clonality: monoclonal
Clone: 11C175
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse