Anti-Protocadherin alpha C2 Mouse Monoclonal Antibody [clone: 9G424]
Lieferant: United States Biological
USBIP9119-10EA 601 EUR
USBIP9119-10
Anti-Protocadherin alpha C2 Mouse Monoclonal Antibody [clone: 9G424]
Antikörper
Type: Primary
Antigen: Protocadherin alpha C2
Clonality: monoclonal
Clone: 9G424
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human