Anti-PCDHA6 Mouse Monoclonal Antibody [clone: 9G425]
Lieferant: United States Biological
USBIP9119-15EA 601 EUR
USBIP9119-15
Anti-PCDHA6 Mouse Monoclonal Antibody [clone: 9G425]
Antikörper
Type: Primary
Antigen: PCDHA6 (protocadherin alpha 6)
Clonality: monoclonal
Clone: 9G425
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human