Anti-PCDHA7 Mouse Polyclonal Antibody

Lieferant: United States Biological
USBIP9119-16EA 555 EUR
USBIP9119-16
Anti-PCDHA7 Mouse Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PCDHA7 (protocadherin alpha 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR