Anti-PCDH11Y Goat Polyclonal Antibody

Lieferant: United States Biological
USBIP9119-21EA 420 EUR
USBIP9119-21
Anti-PCDH11Y Goat Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1: 8000

Type: Primary
Antigen: PCDH11Y (protocadherin 11 Y-linked)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR