Anti-RET Mouse Monoclonal Antibody (Carboxyfluorescein) [clone: 8H101]
Lieferant: United States Biological
USBIR1640-02FEA 525 EUR
USBIR1640-02F
Anti-RET Mouse Monoclonal Antibody (Carboxyfluorescein) [clone: 8H101]
Antikörper
Type: Primary
Antigen: RET (ret proto-oncogene)
Clonality: monoclonal
Clone: 8H101
Conjugation: Carboxyfluorescein
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse