Anti-RET Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 8H101]

Lieferant: United States Biological
USBIR1640-02GEA 606 EUR
USBIR1640-02G
Anti-RET Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 8H101]
Antikörper
Do not freeze!

Type: Primary
Antigen: RET (ret proto-oncogene)
Clonality: monoclonal
Clone: 8H101
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR