Anti-Retinal Pigment Epithelium Specific Protein Mouse Monoclonal Antibody [clone: 8B99]

Lieferant: United States Biological
USBIR1659-70AEA 608 EUR
USBIR1659-70A
Anti-Retinal Pigment Epithelium Specific Protein Mouse Monoclonal Antibody [clone: 8B99]
Antikörper
Recommended dilution: Western Blot: 1:500-1:1000; Immunochemistry: 1:250; Immunofluorescence: 1:10-1:500; Flow Cytometry: 1:10-1:500

Type: Primary
Antigen: Retinal Pigment Epithelium Specific Protein
Clonality: monoclonal
Clone: 8B99
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Bovine, Dog, Human, Mouse, Pig, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR