Anti-DCLRE1A Goat polyclonal antibody

Lieferant: United States Biological
USBIS1014-95PEA 633 EUR
USBIS1014-95P
Anti-DCLRE1A Goat polyclonal antibody
Antikörper
Recommended dilution: ELISA: 1:8000; Immunochemistry: 2.5 ?g/ml

Type: Primary
Antigen: DCLRE1A (DNA cross-link repair 1A)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR