Anti-SNRPD1 Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBIS1014-96EA 587 EUR
USBIS1014-96
Anti-SNRPD1 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:50-1:100

Type: Primary
Antigen: SNRPD1 (small nuclear ribonucleoprotein D1 polypeptide 16kDa)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR