Anti-SNRPA Mouse Monoclonal Antibody [clone: 3F9-1F7]
Lieferant: United States Biological
USBIS1014-96G1EA 703 EUR
USBIS1014-96G1
Anti-SNRPA Mouse Monoclonal Antibody [clone: 3F9-1F7]
Antikörper
Recommended dilution: Immunofluorescence: 10 ?g/ml; Immunochemistry: 5 ?g/ml
Type: Primary
Antigen: SNRPA (small nuclear ribonucleoprotein polypeptide A)
Clonality: Monoclonal
Clone: 3F9-1F7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human