Anti-SNX2 Chicken Polyclonal Antibody

Lieferant: United States Biological
USBIS1014-97BEA 729 EUR
USBIS1014-97B
Anti-SNX2 Chicken Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 1:1000-1:2000

Type: Primary
Antigen: SNX2 (sorting nexin 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR