Anti-SRD5A2 Goat Polyclonal Antibody

Lieferant: United States Biological
USBIS6505-08QEA 551 EUR
USBIS6505-08Q
Anti-SRD5A2 Goat Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:8000; Western Blot: 0. 3?g/ml

Type: Primary
Antigen: SRD5A2 (steroid-5-alpha-reductase, alpha polypeptide 2 (3-oxo-5 alpha-steroid delta 4-dehydrogenase alpha 2))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR