Anti-TRIM21 Goat Polyclonal Antibody

Lieferant: United States Biological
USBIS6600-66CEA 633 EUR
USBIS6600-66C
Anti-TRIM21 Goat Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:32000; Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: TRIM21 (tripartite motif containing 21)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR