Anti-SSX2IP Goat Polyclonal Antibody

Lieferant: United States Biological
USBIS6600-94CEA 516 EUR
USBIS6600-94C
Anti-SSX2IP Goat Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:16000

Type: Primary
Antigen: SSX2IP (synovial sarcoma, X breakpoint 2 interacting protein)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Dog, Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR