Anti-S Tag Mouse Monoclonal Antibody [clone: 10F400]
Lieferant: United States Biological
USBIS6609-95QEA 439 EUR
USBIS6609-95Q
Anti-S Tag Mouse Monoclonal Antibody [clone: 10F400]
Antikörper
Recommended dilution: Flow Cytometry: 1:5-1:10;
Type: Tag
Antigen: S Tag (S tag)
Clonality: Monoclonal
Clone: 10F400
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen