Anti-S Tag Goat Polyclonal Antibody (FITC (Fluorescein))
Lieferant: United States Biological
USBIS6609-96AEA 627 EUR
USBIS6609-96A
Anti-S Tag Goat Polyclonal Antibody (FITC (Fluorescein))
Antikörper
Type: Tag
Antigen: S Tag (S tag)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Goat
Isotype:
Reactivity:
Bestellen