Anti-STAG3 Goat Polyclonal Antibody

Lieferant: United States Biological
USBIS6610-12EA 420 EUR
USBIS6610-12
Anti-STAG3 Goat Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:16000

Type: Primary
Antigen: STAG3 (stromal antigen 3)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR