Anti-STEAP1 Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBIS7500-04EEA 633 EUR
USBIS7500-04E
Anti-STEAP1 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 2?g/ml; Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: STEAP1 (six transmembrane epithelial antigen of the prostate 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR