Anti-Verotoxin I, A/B Subunits Mouse Monoclonal Antibody [clone: 9L400]

Lieferant: United States Biological
USBIV2121-40EA 1030 EUR
USBIV2121-40
Anti-Verotoxin I, A/B Subunits Mouse Monoclonal Antibody [clone: 9L400]
Antikörper
Recommended dilution: ELISA: 20 ?g/ml

Type: Primary
Antigen: Verotoxin I, A/B Subunits
Clonality: monoclonal
Clone: 9L400
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR