Anti-VENTX Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBIV2121-48QEA 450 EUR
USBIV2121-48Q
Anti-VENTX Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 0.5-4?g/ml

Type: Primary
Antigen: VENTX (VENT homeobox)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Xenopus laevis
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR