Anti-VENTX Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBIV2121-48REA 633 EUR
USBIV2121-48R
Anti-VENTX Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 1-2?g/ml; Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: VENTX (VENT homeobox)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR