Anti-Verotoxin I-B subunit Mouse Monoclonal Antibody [clone: 9G300]

Lieferant: United States Biological
USBIV2121-60BEA 782 EUR
USBIV2121-60B
Anti-Verotoxin I-B subunit Mouse Monoclonal Antibody [clone: 9G300]
Antikörper
Recommended dilution: Western Blot: 1:10

Type: Primary
Antigen: Verotoxin I-B subunit
Clonality: monoclonal
Clone: 9G300
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR