Anti-Verotoxin II-B subunit Mouse Monoclonal Antibody [clone: 5E458]

Lieferant: United States Biological
USBIV2121-70AEA 479 EUR
USBIV2121-70A
Anti-Verotoxin II-B subunit Mouse Monoclonal Antibody [clone: 5E458]
Antikörper
Recommended dilution: ELISA: 1:20-1:200

Type: Primary
Antigen: Verotoxin II-B subunit
Clonality: monoclonal
Clone: 5E458
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR