Anti-Verotoxin II-B subunit Mouse Monoclonal Antibody [clone: 9L402]

Lieferant: United States Biological
USBIV2121-70BEA 1220 EUR
USBIV2121-70B
Anti-Verotoxin II-B subunit Mouse Monoclonal Antibody [clone: 9L402]
Antikörper
Recommended dilution: ELISA: 10 ?g/ml; ELISA: 1 ?g/ml; Western Blot: 1 ?g/ml

Type: Primary
Antigen: Verotoxin II-B subunit
Clonality: monoclonal
Clone: 9L402
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR