Anti-VTI1B Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Lieferant: United States Biological
USBIV2121-79-APCEA 852 EUR
USBIV2121-79-APC
Anti-VTI1B Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-1:500
Do not freeze!

Type: Primary
Antigen: VTI1B (vesicle transport through interaction with t-SNAREs 1B)
Clonality: polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR