Anti-SLC18A3 Guinea pig Polyclonal Antibody

Lieferant: United States Biological
USBIV2121-85AEA 998 EUR
USBIV2121-85A
Anti-SLC18A3 Guinea pig Polyclonal Antibody
Antikörper
Recommended dilution: Immunochemistry: 1:5000-1:10000

Type: Primary
Antigen: SLC18A3 (solute carrier family 18 (vesicular acetylcholine transporter), member 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Guinea pig
Isotype: IgG
Reactivity: Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR