Goat Anti-Rabbit IgG Polyclonal Antibody (DyLight® 550)

IMMRIR2187EA 101 EUR
IMMRIR2187
Goat Anti-Rabbit IgG Polyclonal Antibody (DyLight® 550)
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 550
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rabbit
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR