This radiometer measures the UV radiation at specific wavelength (254, 312 or 365 nm), designed for an accurate and direct measurement.
It includes a silicone photo-electric sensor for a direct measurement of the UV intensity and thus do not convert UV into visible light.
Highly accurate and user friendly radiometer to measure the UV radiation dosage according to two parameters: Time (min or sec) or energy (J/cm²). It is plugable to interchangeable sensors – See accessories.
Bestellinformation: Sensors must be ordered separately.