Dispensers for cleanroom wipes
Lieferant: AK
AKAAAK-109-LEA 174 EUR
AKAAAK-109-L
Dispensers for cleanroom wipes
Wischtuch-Spender
This wipe holder with slanted interior and front cut out provides you with fingertip access to cleanroom wipes at all times. Made from PETG, clear, thickness: 3 mm.
  • Resistant to IPA solution
  • Made of high impact PETG
  • Wall-mountable of free-standing