Anti-GNAI2 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA318889EA 590 EUR
ORIGTA318889
Anti-GNAI2 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 0.5-4?g/ml; Immunoprecipitation: 10-20?g/ml

Type: Primary
Antigen: GNAI2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR