Anti-Hdac7 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA318926EA 618 EUR
ORIGTA318926
Anti-Hdac7 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 0.5-4 ?g/ml; Immunochemistry: 20-40?g/ml; Immunoprecipitation: 20-40?g/ml

Type: Primary
Antigen: Hdac7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR