Anti-DPPA4 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA318970EA 582 EUR
ORIGTA318970
Anti-DPPA4 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 0.5-4 ?g/ml

Type: Primary
Antigen: DPPA4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR