Anti-Scd Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA318988EA 590 EUR
ORIGTA318988
Anti-Scd Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 0.5-4 ?g/ml

Type: Primary
Antigen: Scd
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Hamster, Human, Mouse, Pig, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR