Anti-Il2 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA319037EA 578 EUR
ORIGTA319037
Anti-Il2 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1-4 ?g/ml

Type: Primary
Antigen: Il2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR