Anti-Cxcl12 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA319083EA 689 EUR
ORIGTA319083
Anti-Cxcl12 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:1000-2500

Type: Primary
Antigen: Cxcl12
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR