Anti-VIM Mouse monoclonal Antibody (Biotin) [clone: 1A9]

Lieferant: OriGene
ORIGTA700389EA 634 EUR
ORIGTA700389
Anti-VIM Mouse monoclonal Antibody (Biotin) [clone: 1A9]
Antikörper
Recommended Dilutions: 1:250-1:1000

Type: Primary
Antigen: VIM
Clonality: monoclonal
Clone: 1A9
Conjugation: Biotin
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR