Anti-CST2 Mouse monoclonal Antibody (Biotin) [clone: 1D6]

Lieferant: OriGene
ORIGTA700406EA 642 EUR
ORIGTA700406
Anti-CST2 Mouse monoclonal Antibody (Biotin) [clone: 1D6]
Antikörper
Type: Primary
Antigen: CST2
Clonality: monoclonal
Clone: 1D6
Conjugation: Biotin
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR