Anti-MADCAM1 Mouse Monoclonal Antibody [clone: 1B7]

Lieferant: OriGene
ORIGTA507035EA 581 EUR
ORIGTA507035
Anti-MADCAM1 Mouse Monoclonal Antibody [clone: 1B7]
Antikörper
Recommended Dilutions: Western Blot: 1:400-4000

Type: Primary
Antigen: MADCAM1
Clonality: Monoclonal
Clone: 1B7
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR