Anti-SHBG Mouse Monoclonal Antibody [clone: 1G2]

Lieferant: OriGene
ORIGTA507184EA 603 EUR
ORIGTA507184
Anti-SHBG Mouse Monoclonal Antibody [clone: 1G2]
Antikörper
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: SHBG
Clonality: Monoclonal
Clone: 1G2
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR