Anti-TP63 Mouse Monoclonal Antibody [clone: 4F3]

Lieferant: OriGene
ORIGTA600388EA 603 EUR
ORIGTA600388
Anti-TP63 Mouse Monoclonal Antibody [clone: 4F3]
Antikörper
Recommended Dilutions: 1:100-1:1000

Type: Primary
Antigen: TP63
Clonality: Monoclonal
Clone: 4F3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR