Anti-ALDH1L1 Mouse Monoclonal Antibody [clone: 4C11]

Lieferant: OriGene
ORIGTA600396EA 603 EUR
ORIGTA600396
Anti-ALDH1L1 Mouse Monoclonal Antibody [clone: 4C11]
Antikörper
Type: Primary
Antigen: ALDH1L1
Clonality: Monoclonal
Clone: 4C11
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR