Anti-EFNA2 Mouse Monoclonal Antibody [clone: 2D6]

Lieferant: OriGene
ORIGTA600409EA 603 EUR
ORIGTA600409
Anti-EFNA2 Mouse Monoclonal Antibody [clone: 2D6]
Antikörper
Type: Primary
Antigen: EFNA2
Clonality: Monoclonal
Clone: 2D6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR