Anti-KRT7 Mouse Monoclonal Antibody [clone: 1C5]

Lieferant: OriGene
ORIGTA802932EA 603 EUR
ORIGTA802932
Anti-KRT7 Mouse Monoclonal Antibody [clone: 1C5]
Antikörper
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:150

Type: Primary
Antigen: KRT7
Clonality: Monoclonal
Clone: 1C5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR