Anti-AVPI1 Mouse Monoclonal Antibody [clone: 6C5]

Lieferant: OriGene
ORIGTA802991EA 603 EUR
ORIGTA802991
Anti-AVPI1 Mouse Monoclonal Antibody [clone: 6C5]
Antikörper
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: AVPI1
Clonality: Monoclonal
Clone: 6C5
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR