Anti-MRPS11 Mouse Monoclonal Antibody [clone: 2D1]

Lieferant: OriGene
ORIGTA802879EA 603 EUR
ORIGTA802879
Anti-MRPS11 Mouse Monoclonal Antibody [clone: 2D1]
Antikörper
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: MRPS11
Clonality: Monoclonal
Clone: 2D1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR