Anti-FRK Mouse Monoclonal Antibody [clone: 7H3]

Lieferant: OriGene
ORIGCF500527EA 749 EUR
ORIGCF500527
Anti-FRK Mouse Monoclonal Antibody [clone: 7H3]
Antikörper
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:50; Flow Cytometry: 1:100

Type: Primary
Antigen: FRK
Clonality: Monoclonal
Clone: 7H3
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR