Anti-BUB1B Mouse Monoclonal Antibody [clone: 5D9]

Lieferant: OriGene
ORIGCF500533EA 749 EUR
ORIGCF500533
Anti-BUB1B Mouse Monoclonal Antibody [clone: 5D9]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50 Immunofluorescence: 1:100

Type: Primary
Antigen: BUB1B
Clonality: Monoclonal
Clone: 5D9
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR